Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ampd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138876.1 | NP_620231.1 | 12 | 1633 | ||
XM_006233060.2 | XP_006233122.1 | 11 | 1710 | |||
XM_008761369.1 | XP_008759591.1 | 11 | 1633 | |||
GenBank mRNA | FQ214856.1 | - | 5 | 732 | ||
FQ216456.1 | - | 2 | 184 | |||
FQ224901.1 | - | 2 | 121 | |||
J02811.1 | - | 12 | 1633 |