Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139110.1 | NP_620810.1 | 2 | 270 | ||
XM_006244594.2 | XP_006244656.1 | 2 | 485 | |||
XM_006244597.2 | XP_006244659.1 | 2 | 484 | |||
XM_008766832.1 | XP_008765054.1 | 2 | 487 | |||
XM_008766833.1 | XP_008765055.1 | 2 | 412 | |||
XM_008766834.1 | XP_008765056.1 | 2 | 488 | |||
XM_008766835.1 | XP_008765057.1 | 2 | 486 | |||
GenBank mRNA | AB019120.1 | - | 2 | 270 |