Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139110.1 | NP_620810.1 | 3 | 321 | ||
XM_006244594.2 | XP_006244656.1 | 3 | 536 | |||
XM_006244597.2 | XP_006244659.1 | 3 | 535 | |||
XM_008766832.1 | XP_008765054.1 | 3 | 538 | |||
XM_008766833.1 | XP_008765055.1 | 3 | 463 | |||
XM_008766834.1 | XP_008765056.1 | 3 | 539 | |||
XM_008766835.1 | XP_008765057.1 | 3 | 537 | |||
GenBank mRNA | AB019120.1 | - | 3 | 321 |
Set Membership: |
G-protein coupled receptor |