Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145090.3 | NP_659558.1 | 9, 10 | 969 | ||
XM_006235731.2 | XP_006235793.1 | 9, 10 | 1069 | |||
XM_006235732.1 | XP_006235794.1 | 9, 10 | 1010 | |||
XM_006235733.2 | XP_006235795.1 | 8, 9 | 1278 | |||
XM_006235734.2 | XP_006235796.1 | 8, 9 | 1277 | |||
GenBank mRNA | BC070895.1 | - | 9, 10 | 950 | 7099840 | |
CV120035.1 | - | 1, 2 | 60 | |||
DQ167810.1 | - | 7, 8 | 686 | |||
DQ167811.1 | - | 7, 8 | 686 | |||
FQ222295.1 | - | 9, 10 | 992 | |||
FQ232032.1 | - | 9, 10 | 990 | |||
FQ233265.1 | - | 9, 10 | 990 | |||
U35776.1 | - | 9, 10 | 969 |