Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153312.2 | NP_695224.2 | 1261 | |||
GenBank mRNA | BC089765.1 | - | 1266 | 7372112 | ||
FQ210941.1 | - | 53 | ||||
M13646.1 | - | 1261 | ||||
U09742.1 | - | 1263 | ||||
X79320.1 | - | 1263 |