Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271058.1 | NP_001257987.1 | 2, 3 | 242 | ||
NM_001271059.1 | NP_001257988.1 | 3, 4 | 286 | |||
NM_001271060.1 | NP_001257989.1 | 2, 3 | 277 | |||
NM_001271061.1 | NP_001257990.1 | 2, 3 | 211 | |||
NM_001271062.1 | NP_001257991.1 | 2, 3 | 320 | |||
NM_173096.3 | NP_775119.2 | 2, 3 | 400 | |||
XM_006248146.2 | XP_006248208.1 | 3, 4 | 466 | |||
XM_006248149.2 | XP_006248211.1 | 3, 4 | 509 | |||
XM_008768555.1 | XP_008766777.1 | 2, 3 | 2402 | |||
XM_008768556.1 | XP_008766778.1 | 2, 3 | 2375 | |||
XM_008768557.1 | XP_008766779.1 | 2, 3 | 2310 | |||
GenBank mRNA | BC099784.1 | - | 2, 3 | 388 | 7933587 | |
FQ226931.1 | - | 2, 3 | 277 | |||
FQ230542.1 | - | 2, 3 | 289 | |||
FQ231581.1 | - | 3, 4 | 354 | |||
X52711.1 | - | 1, 2 | 194 |