Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177421.3 | NP_803156.2 | 10 | 2051 | ||
XM_006251965.1 | XP_006252027.1 | 10 | 2689 | |||
GenBank mRNA | AB040056.1 | - | 10 | 2059 | ||
BC099072.1 | - | 10 | 2051 | 7312276 | ||
FQ212273.1 | - | 10 | 2053 |