Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182738.1 | NP_877402.1 | 4 | 378 | ||
XM_008759745.1 | XP_008757967.1 | 5 | 556 | |||
XM_008759746.1 | XP_008757968.1 | 5 | 526 | |||
GenBank mRNA | AB086189.1 | - | 4 | 378 |