Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183054.1 | NP_898877.1 | 1 | 185 | ||
XM_006232631.2 | XP_006232693.1 | 1 | 446 | |||
XM_006232633.2 | XP_006232695.1 | 1 | 673 | |||
GenBank mRNA | AY129072.1 | - | 1 | 185 | ||
BC079365.1 | - | 1 | 167 | 7109321 |