Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201415.1 | NP_958818.1 | 7 | 584 | ||
GenBank mRNA | BC098644.1 | - | 6 | 569 | 7129700 | |
FQ211037.1 | - | 7 | 590 | |||
FQ211890.1 | - | 7 | 590 | |||
FQ221050.1 | - | 7 | 590 | |||
FQ221111.1 | - | 7 | 590 | |||
FQ221445.1 | - | 7 | 590 | |||
FQ221630.1 | - | 7 | 592 | |||
FQ222545.1 | - | 7 | 590 | |||
FQ222824.1 | - | 7 | 590 | |||
FQ223312.1 | - | 7 | 590 | |||
FQ223528.1 | - | 7 | 590 | |||
FQ223728.1 | - | 7 | 589 | |||
FQ224659.1 | - | 7 | 590 | |||
FQ228520.1 | - | 7 | 590 | |||
FQ228658.1 | - | 4 | 417 | |||
FQ229921.1 | - | 7 | 587 | |||
M60478.1 | - | 4 | 364 | |||
X58389.1 | - | 7 | 585 | |||
X60212.1 | - | 7 | 584 |
Gene Symbol: Hnrnph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256376.2 | XP_006256438.1 | 1 | 1018 | ||
XM_006256377.2 | XP_006256439.1 | 1 | 1018 | |||
XM_006256378.2 | XP_006256440.1 | 1 | 1018 | |||
XM_006256379.2 | XP_006256441.1 | 1 | 1018 |
Gene Symbol: LOC691392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001075994.5 | XP_001075994.3 | 2 | 677 |