Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201416.1 | NP_958819.1 | 1 | 155 | ||
XM_006255758.2 | XP_006255820.1 | 2 | 818 | |||
XM_006255759.2 | XP_006255821.1 | 2 | 819 | |||
XM_006255760.2 | XP_006255822.1 | 2 | 817 | |||
XM_006255761.2 | XP_006255823.1 | 2 | 329 | |||
XM_008772603.1 | XP_008770825.1 | 2 | 237 | |||
GenBank mRNA | AY389807.1 | - | 1 | 155 | ||
AY389808.1 | - | 1 | 155 |