Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rxfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201417.1 | NP_958820.1 | 3 | 275 | ||
XM_006232508.2 | XP_006232570.1 | 3 | 415 | |||
XM_006232510.2 | XP_006232572.1 | 1 | 72 | |||
GenBank mRNA | AY509976.1 | - | 3 | 275 |