Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201991.1 | NP_973720.1 | 6 | 669 | ||
GenBank mRNA | AY513752.1 | - | 6 | 669 |