Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203334.1 | NP_976079.1 | 6 | 1358 | ||
XM_008759310.1 | XP_008757532.1 | 6 | 2169 | |||
XM_008759311.1 | XP_008757533.1 | 7 | 1367 | |||
GenBank mRNA | AY547309.1 | - | 6 | 1250 | ||
L21672.2 | - | 6 | 1358 |