Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145044.1 | NP_001138516.1 | 1 | 275 | ||
NM_013272.3 | NP_037404.2 | 1 | 275 | |||
XM_005254889.1 | XP_005254946.1 | 1 | 164 | |||
GenBank mRNA | AB031050.2 | - | 1 | 267 | ||
AF187816.1 | - | 1 | 74 | |||
AF205074.1 | - | 1 | 263 | |||
AK290872.1 | - | 1 | 113 | |||
BC000585.2 | - | 1 | 127 | 3163826 |