Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RACGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126103.2 | NP_001119575.1 | 5 | 467 | ||
NM_001126104.2 | NP_001119576.1 | 3 | 347 | |||
NM_001319999.1 | NP_001306928.1 | 3 | 277 | |||
NM_001320000.1 | NP_001306929.1 | 4 | 351 | |||
NM_001320001.1 | NP_001306930.1 | 3 | 343 | |||
NM_001320002.1 | NP_001306931.1 | 3 | 302 | |||
NM_001320003.1 | NP_001306932.1 | 3 | 273 | |||
NM_001320004.1 | NP_001306933.1 | 3 | 224 | |||
NM_001320005.1 | NP_001306934.1 | 2 | 188 | |||
NM_001320006.1 | NP_001306935.1 | 4 | 347 | |||
NM_013277.4 | NP_037409.2 | 5 | 456 | |||
XM_006719359.1 | XP_006719422.1 | 3 | 175 | |||
XM_011538238.1 | XP_011536540.1 | 3 | 132 | |||
XM_017019220.1 | XP_016874709.1 | 3 | 449 | |||
GenBank mRNA | AB030251.1 | - | 3 | 183 | ||
AK000733.1 | - | 3 | 190 | |||
AK225901.1 | - | 2 | 118 | |||
AK315785.1 | - | 5 | 404 | |||
AL136794.1 | - | 5 | 337 | |||
BC024144.2 | - | 3 | 168 | |||
BC032754.1 | - | 3 | 174 | 5583315 | ||
BU940828.1 | - | 4 | 241 | |||
CR533565.1 | - | 2 | 113 | |||
DA051287.1 | - | 3 | 181 | |||
DA441098.1 | - | 3 | 245 | |||
DA502516.1 | - | 3 | 159 | |||
DA812983.1 | - | 4 | 255 | |||
DB076571.1 | - | 3 | 244 | |||
DB084617.1 | - | 3 | 225 | |||
DB090068.1 | - | 2 | 125 | |||
DB164810.1 | - | 3 | 160 |