Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207586.2 | NP_997469.1 | 6 | 524 | ||
GenBank mRNA | AY486074.1 | - | 6 | 526 | ||
KC529649.1 | - | 5 | 315 |