Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013281.3 | NP_037413.1 | 2 | 487 | ||
NM_198391.2 | NP_938205.1 | 3 | 681 | |||
XM_005260682.4 | XP_005260739.1 | 3 | 1199 | |||
XM_011529204.2 | XP_011527506.1 | 4 | 1336 | |||
XM_011529205.2 | XP_011527507.1 | 4 | 998 | |||
GenBank mRNA | AB040902.1 | - | 2 | 375 | ||
AF169677.1 | - | 2 | 384 | |||
AK027297.1 | - | 2 | 398 | |||
AK027694.1 | - | 1 | 116 | |||
AK074883.1 | - | 2 | 428 | |||
AK074909.1 | - | 2 | 356 | |||
AL529481.3 | - | 3 | 637 | |||
AY358319.1 | - | 1 | 221 | |||
BC020870.1 | - | 2 | 394 | 4606852 | ||
CD654700.1 | - | 2 | 448 | |||
DA502792.1 | - | 2 | 487 |