Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284331.1 | NP_001271260.1 | 5, 6 | 808 | ||
NM_013284.3 | NP_037416.1 | 5, 6 | 808 | |||
XM_006715692.2 | XP_006715755.1 | 5, 6 | 779 | |||
XM_006715696.2 | XP_006715759.1 | 3, 4 | 509 | |||
XM_011515275.1 | XP_011513577.1 | 5, 6 | 794 | |||
XM_011515279.1 | XP_011513581.1 | 5, 6 | 794 | |||
GenBank mRNA | AF176097.1 | - | 5, 6 | 748 | ||
AJ131891.2 | - | 5, 6 | 751 | |||
AK307634.1 | - | 3, 4 | 481 | |||
BC062590.1 | - | 5, 6 | 732 | 5215523 |