Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207601.1 | NP_997484.1 | 5 | 979 | ||
GenBank mRNA | BC061844.1 | - | 5 | 979 | 5621135 | |
FQ220564.1 | - | 5 | 1018 |