Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hyal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207616.1 | NP_997499.1 | 2 | 123 | ||
GenBank mRNA | AB100600.1 | - | 2 | 123 |