Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005063.4 | NP_005054.3 | 6 | 2633 | ||
GenBank mRNA | AB032261.1 | - | 6 | 2513 | ||
AF097514.1 | - | 6 | 2379 | |||
AF109362.1 | - | 1 | 357 |