Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSK3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146156.1 | NP_001139628.1 | 2 | 1087 | ||
NM_002093.3 | NP_002084.2 | 2 | 1087 | |||
XM_006713610.2 | XP_006713673.1 | 2 | 573 | |||
XM_006713611.2 | XP_006713674.1 | 2 | 573 | |||
GenBank mRNA | AB451356.1 | - | 2 | 104 | ||
AK290897.1 | - | 2 | 460 | |||
BC000251.1 | - | 2 | 336 | 3357620 | ||
BC012760.2 | - | 2 | 1086 | 3637163 | ||
CR536510.1 | - | 2 | 104 | |||
EU302497.1 | - | 2 | 104 | |||
L33801.1 | - | 2 | 143 |