Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTNR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005958.4 | NP_005949.1 | 1, 2 | 386 | ||
GenBank mRNA | BC074946.2 | - | 1, 2 | 202 | 30915497 | |
BC074947.2 | - | 1, 2 | 202 | 30915274 | ||
BC126297.1 | - | 1, 2 | 386 | 8992013 | ||
BC126299.1 | - | 1, 2 | 384 | 8992015 | ||
EU432127.1 | - | 1, 2 | 184 | |||
U14108.1 | - | 1, 2 | 216 |
Set Membership: |
G-protein coupled receptor |