Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013306.4 | NP_037438.2 | 1 | 207 | ||
NM_147777.3 | NP_680086.2 | 1 | 207 | |||
GenBank mRNA | AB209439.1 | - | 1 | 75 | ||
AF001435.1 | - | 1 | 133 | |||
AF175267.2 | - | 1 | 116 | |||
AF175268.1 | - | 1 | 77 | |||
AK315034.1 | - | 1 | 138 | |||
AK315642.1 | - | 1 | 148 | |||
BC009897.2 | - | 1 | 125 | 3510379 | ||
BC012767.2 | - | 1 | 177 | 3678551 | ||
BC014520.2 | - | 1 | 177 | 3678576 | ||
BT006631.1 | - | 1 | 77 | |||
DA079695.1 | - | 1 | 208 |
Gene Symbol: ARL2-SNX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037650.1 | 4 | 731 | |||
GenBank mRNA | AK096447.1 | - | 4 | 665 |