Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003812.3 | NP_003803.1 | 11 | 1299 | ||
XM_005246932.3 | XP_005246989.1 | 11 | 1399 | |||
XM_011512086.2 | XP_011510388.1 | 11 | 1399 | |||
GenBank mRNA | AB009672.1 | - | 11 | 1299 | ||
AJ005580.1 | - | 11 | 1128 | |||
AK129906.1 | - | 7 | 463 | |||
AK296844.1 | - | 11 | 1008 | |||
BC132763.1 | - | 11 | 1279 | 40146785 | ||
BC132765.1 | - | 11 | 1279 | 40146787 |