Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004401.2 | NP_004392.1 | 4 | 583 | ||
NM_213566.1 | NP_998731.1 | 4 | 583 | |||
GenBank mRNA | AF087573.1 | - | 4 | 522 | ||
AF103799.1 | - | 4 | 510 | |||
AK222546.1 | - | 4 | 519 | |||
AK313317.1 | - | 4 | 517 | |||
AU121791.1 | - | 4 | 583 | |||
BC000037.2 | - | 4 | 503 | 3505817 | ||
BC007112.1 | - | 4 | 554 | 4292811 | ||
BC007721.2 | - | 4 | 515 | 4299734 | ||
BT006980.1 | - | 4 | 485 | |||
U91985.1 | - | 4 | 541 |