Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100176.1 | NP_001093646.1 | 3, 4 | 383 | ||
NM_013312.2 | NP_037444.2 | 3, 4 | 383 | |||
XM_005259873.2 | XP_005259930.1 | 3, 4 | 383 | |||
XM_005259874.2 | XP_005259931.1 | 3, 4 | 383 | |||
GenBank mRNA | AF044924.1 | - | 3, 4 | 286 | ||
BC012443.1 | - | 3, 4 | 306 | 3859634 | ||
DB474423.1 | - | 3, 4 | 383 |