Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005290.3 | NP_005281.1 | 1 | 580 | ||
GenBank mRNA | AK313375.1 | - | 1 | 580 | ||
BC069437.1 | - | 1 | 445 | 7262234 | ||
BC101779.1 | - | 1 | 471 | 8069285 | ||
BC101781.1 | - | 1 | 468 | 8069287 |
Set Membership: |
G-protein coupled receptor |