Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YPEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013313.4 | NP_037445.1 | 5 | 699 | ||
NR_130910.1 | 5 | 1083 | ||||
GenBank mRNA | AB098734.1 | - | 5 | 699 | ||
AB098735.1 | - | 5 | 699 | |||
AF060862.1 | - | 4 | 399 | |||
AF305069.1 | - | 4 | 459 | |||
BC034486.1 | - | 5 | 557 | 4826997 | ||
BI559478.1 | - | 5 | 578 | |||
CR456376.2 | - | 4 | 346 | |||
CR933630.1 | - | 4 | 4802 |