Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other VRK2 SIRNA Assays ›
Gene Symbol:
VRK2
FANCL
Gene Name:
vaccinia related kinase 2
Fanconi anemia complementation group L
Gene Aliases:
-
FAAP43, PHF9, POG
Chromosome Location:
Chr.2: 57907651 - 58163991 on Build GRCh38
Chromosome Location:
Chr.2: 58159243 - 58241380 on Build GRCh38
UniGene ID:
Hs.715298
Hs.631890
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
202109
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: VRK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130480.2
NP_001123952.1
13
1486
NM_001130481.2
NP_001123953.1
13
1328
NM_001130482.2
NP_001123954.1
13
1432
NM_001130483.2
NP_001123955.1
14
1535
NM_001288836.1
NP_001275765.1
14
1623
NM_001288837.1
NP_001275766.1
16
2046
NM_001288838.1
NP_001275767.1
16
1977
NM_001288839.1
NP_001275768.1
12
1463
NM_006296.6
NP_006287.2
13
1533
XM_005264540.4
XP_005264597.1
13
1546
XM_006712090.3
XP_006712153.1
14
1385
XM_006712091.3
XP_006712154.1
14
1630
XM_006712092.3
XP_006712155.1
14
1718
XM_006712093.3
XP_006712156.1
11
1423
XM_011533092.2
XP_011531394.1
13
1330
XM_017004858.1
XP_016860347.1
14
1530
XM_017004859.1
XP_016860348.1
14
2208
XM_017004860.1
XP_016860349.1
13
2138
XM_017004861.1
XP_016860350.1
11
1237
XM_017004862.1
XP_016860351.1
12
2050
XM_017004863.1
XP_016860352.1
13
1630
XM_017004864.1
XP_016860353.1
15
1762
XM_017004865.1
XP_016860354.1
15
1581
XM_017004866.1
XP_016860355.1
15
1630
GenBank mRNA
AB000450.1
-
13
1431
AJ512204.2
-
14
1475
AK058199.1
-
13
1388
AK223540.1
-
13
1388
AK292727.1
-
16
1963
AK296611.1
-
14
1608
AK311205.1
-
4
417
AY228367.1
-
13
1328
AY228368.1
-
13
1463
AY228369.1
-
13
1361
AY228370.1
-
14
1553
AY228371.1
-
12
1393
AY228372.1
-
14
1465
BC027854.1
-
13
1484
5221315
Gene Symbol: FANCL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
BQ574618.1
-
1
241
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
regulation of cell shape
viral process
peptidyl-serine phosphorylation
cellular response to oxidative stress
regulation of MAPK cascade
protein autophosphorylation
regulation of interleukin-1-mediated signaling pathway
protein serine/threonine kinase activity
protein binding
ATP binding
protein kinase binding
protein domain specific binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0