Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318014.1 | NP_001304943.1 | 4 | 3061 | ||
NM_001318022.1 | NP_001304951.1 | 4 | 2944 | |||
NM_030883.4 | NP_112145.1 | 4 | 2948 | |||
GenBank mRNA | AJ459850.1 | - | 1 | 97 | ||
BC048991.2 | - | 3 | 2896 | 5271584 |