Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145059.2 | NP_659496.2 | 17 | 2136 | ||
XM_006721162.1 | XP_006721225.1 | 16 | 2066 | |||
XM_011522928.1 | XP_011521230.1 | 17 | 2314 | |||
XM_011522929.1 | XP_011521231.1 | 17 | 2179 | |||
XM_011522930.2 | XP_011521232.1 | 17 | 2147 | |||
XM_011522931.2 | XP_011521233.1 | 17 | 2147 | |||
XM_017023012.1 | XP_016878501.1 | 17 | 2163 | |||
XM_017023013.1 | XP_016878502.1 | 14 | 1959 | |||
XM_017023014.1 | XP_016878503.1 | 12 | 2091 | |||
GenBank mRNA | AJ441184.1 | - | 17 | 2135 | ||
AK056456.1 | - | 17 | 2135 | |||
AK092025.1 | - | 9 | 1285 | |||
AK096727.1 | - | 9 | 1342 | |||
AK128387.1 | - | 17 | 2232 | |||
AK315590.1 | - | 17 | 2139 | |||
BC013735.1 | - | 15 | 1922 | |||
BC032542.1 | - | 17 | 2142 | 5534376 |