Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000741.3 | NP_000732.2 | 1 | 320 | ||
GenBank mRNA | AF385590.1 | - | 1 | 269 | ||
AF498918.1 | - | 1 | 269 | |||
BC095546.1 | - | 1 | 270 | |||
BC119775.1 | - | 1 | 268 | |||
BC119817.1 | - | 1 | 268 | |||
BC137535.1 | - | 1 | 320 | 9021535 |
Set Membership: |
G-protein coupled receptor |