Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000861.3 | NP_000852.1 | 3 | 3163 | ||
NM_001098211.1 | NP_001091681.1 | 2 | 3084 | |||
NM_001098212.1 | NP_001091682.1 | 2 | 3034 | |||
NM_001098213.1 | NP_001091683.1 | 2 | 3314 | |||
XM_011533652.1 | XP_011531954.1 | 4 | 3451 | |||
XM_011533653.2 | XP_011531955.1 | 2 | 3280 | |||
XM_017006283.1 | XP_016861772.1 | 2 | 3234 | |||
XM_017006284.1 | XP_016861773.1 | 2 | 3273 | |||
GenBank mRNA | BC060802.1 | - | 3 | 3165 | 30340368 |
Set Membership: |
G-protein coupled receptor |