Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCRTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001525.2 | NP_001516.2 | 9 | 1580 | ||
XM_017001105.1 | XP_016856594.1 | 9 | 1848 | |||
XM_017001106.1 | XP_016856595.1 | 9 | 1504 | |||
GenBank mRNA | AF041243.1 | - | 7 | 1348 | ||
AK290521.1 | - | 9 | 1580 | |||
BC074796.2 | - | 7 | 1237 | 30915259 | ||
DA803650.1 | - | 1 | 139 |
Set Membership: |
G-protein coupled receptor |