Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128325.2 | NP_001121797.1 | 6 | 1061 | ||
NM_001199021.1 | NP_001185950.1 | 7 | 1415 | |||
NM_012445.3 | NP_036577.1 | 5 | 1119 | |||
GenBank mRNA | AB027466.1 | - | 5 | 1065 | ||
AK024499.1 | - | 2 | 803 | |||
AK074618.1 | - | 5 | 926 | |||
AK074770.1 | - | 5 | 1057 | |||
AK130164.1 | - | 5 | 837 | |||
AY358948.1 | - | 5 | 1064 | |||
BC002707.2 | - | 5 | 849 | 3630467 | ||
BC036341.1 | - | 5 | 1058 | 5183118 |