Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256732.1 | NP_001243661.1 | 6, 7 | 638 | ||
NM_001256734.1 | NP_001243663.1 | 5, 6 | 548 | |||
NM_001256735.1 | NP_001243664.1 | 5, 6 | 548 | |||
NM_001256736.1 | NP_001243665.1 | 5, 6 | 548 | |||
NM_012446.3 | NP_036578.2 | 6, 7 | 638 | |||
XM_011543297.2 | XP_011541599.1 | 6, 7 | 553 | |||
XM_017009304.1 | XP_016864793.1 | 6, 7 | 456 | |||
XM_017009305.1 | XP_016864794.1 | 7, 8 | 569 | |||
XM_017009306.1 | XP_016864795.1 | 6, 7 | 553 | |||
XM_017009307.1 | XP_016864796.1 | 5, 6 | 366 | |||
XM_017009308.1 | XP_016864797.1 | 7, 8 | 717 | |||
XM_017009309.1 | XP_016864798.1 | 6, 7 | 781 | |||
XM_017009310.1 | XP_016864799.1 | 6, 7 | 929 | |||
GenBank mRNA | AF077048.1 | - | 6, 7 | 475 | ||
AF161465.1 | - | 6, 7 | 468 | |||
AK074537.1 | - | 6, 7 | 433 | |||
AK294864.1 | - | 5, 6 | 343 | |||
AK297494.1 | - | 6, 7 | 346 | |||
AK299858.1 | - | 5, 6 | 392 | |||
AK312337.1 | - | 6, 7 | 482 | |||
AL080076.1 | - | 6, 7 | 486 | |||
BC017020.1 | - | 6, 7 | 486 | 3913337 | ||
BI517313.1 | - | 6, 7 | 638 | |||
CR936787.1 | - | 5, 6 | 393 |