Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAS2R8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023918.1 | NP_076407.1 | 1 | 625 | ||
GenBank mRNA | BC069084.1 | - | 1 | 625 | ||
BC069551.1 | - | 1 | 625 | 7262241 | ||
BC096701.1 | - | 1 | 625 | 40015728 | ||
BC096735.1 | - | 1 | 625 | 40015730 | ||
BC098167.1 | - | 1 | 625 | 40015729 | ||
BC098353.1 | - | 1 | 625 | 40015727 |