Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002596.3 | NP_002587.2 | 16 | 2100 | ||
NM_212502.2 | NP_997667.1 | 16 | 2018 | |||
NM_212503.2 | NP_997668.1 | 16 | 2108 | |||
XM_011509602.2 | XP_011507904.1 | 16 | 2158 | |||
XM_017001423.1 | XP_016856912.1 | 16 | 2253 | |||
GenBank mRNA | AB209307.1 | - | 14 | 2680 | ||
AK126879.1 | - | 11 | 2891 | |||
AK293756.1 | - | 15 | 2382 | |||
AK296320.1 | - | 16 | 2066 | |||
AL161977.1 | - | 7 | 2010 | |||
BC000281.2 | - | 10 | 1185 | |||
BC011526.1 | - | 16 | 1953 | 3855304 | ||
BC040529.1 | - | 7 | 2106 |