Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002596.3 | NP_002587.2 | 16 | 2706 | ||
NM_212502.2 | NP_997667.1 | 16 | 2624 | |||
NM_212503.2 | NP_997668.1 | 16 | 2714 | |||
XM_011509602.2 | XP_011507904.1 | 16 | 2764 | |||
XM_017001423.1 | XP_016856912.1 | 16 | 2859 | |||
GenBank mRNA | AB209307.1 | - | 14 | 3286 | ||
AK126879.1 | - | 11 | 3497 | |||
AL161977.1 | - | 7 | 2616 | |||
BC000281.2 | - | 10 | 1791 | |||
BC011526.1 | - | 16 | 2559 | 3855304 | ||
BC040529.1 | - | 7 | 2712 | |||
BM695474.1 | - | 1 | 13 |