Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031454.1 | NP_113642.1 | 3 | 893 | ||
GenBank mRNA | AY324823.1 | - | 3 | 893 | ||
BC001099.2 | - | 3 | 383 | |||
BC020510.1 | - | 2 | 204 | |||
BC110866.1 | - | 3 | 863 | 6041782 | ||
CR456464.1 | - | 3 | 409 |