Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178863.4 | NP_849194.1 | 4, 5 | 764 | ||
NR_110933.1 | 5 | 1302 | ||||
XM_011545783.2 | XP_011544085.1 | 4, 5 | 833 | |||
XM_011545784.2 | XP_011544086.1 | 5, 6 | 852 | |||
XM_017023104.1 | XP_016878593.1 | 5, 6 | 798 | |||
GenBank mRNA | AF289573.1 | - | 4, 5 | 711 | ||
AF401315.2 | - | 4, 5 | 676 | |||
AK097407.1 | - | 5 | 907 | |||
AK289630.1 | - | 4, 5 | 741 | |||
AY027918.1 | - | 4, 5 | 688 | |||
BC036228.1 | - | 4, 5 | 756 | 5276078 |