Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OBP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288987.1 | NP_001275916.1 | 7 | 941 | ||
NM_014581.3 | NP_055396.1 | 7 | 677 | |||
NR_110242.1 | 8 | 783 | ||||
NR_110243.1 | 6 | 543 | ||||
XM_006717086.3 | XP_006717149.1 | 7 | 745 | |||
XM_017014654.1 | XP_016870143.1 | 8 | 913 | |||
XM_017014655.1 | XP_016870144.1 | 9 | 1388 | |||
GenBank mRNA | AY358981.1 | - | 7 | 670 | ||
BX101545.1 | - | 5 | 398 |