Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIAL4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135789.3 | NP_001129261.2 | 1 | 458 | ||
GenBank mRNA | AK123006.1 | - | 2 | 809 | ||
BC130376.1 | - | 1 | 416 | 40146407 | ||
BC130378.1 | - | 1 | 416 | 40146409 |
Gene Symbol: LOC105371242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011510267.2 | XP_011508569.1 | 1 | 575 |
Gene Symbol: PPIAL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143883.2 | NP_001137355.1 | 1 | 458 | ||
GenBank mRNA | AB084917.1 | - | 1 | 458 |
Gene Symbol: PPIAL4G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123068.1 | NP_001116540.1 | 1 | 416 |