Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278554.1 | NP_001265483.1 | 2 | 478 | ||
NM_001278555.1 | NP_001265484.1 | 2 | 286 | |||
NM_006357.3 | NP_006348.1 | 2 | 517 | |||
NM_182678.2 | NP_872619.1 | 3 | 318 | |||
XM_005246244.2 | XP_005246301.1 | 2 | 474 | |||
XM_011510492.2 | XP_011508794.1 | 2 | 717 | |||
XM_017003166.1 | XP_016858655.1 | 2 | 717 | |||
XM_017003167.1 | XP_016858656.1 | 2 | 711 | |||
GenBank mRNA | AB017644.1 | - | 2 | 160 | ||
AF085362.1 | - | 2 | 218 | |||
AK314145.1 | - | 2 | 488 | |||
BC003554.1 | - | 2 | 200 | 3537280 | ||
BC092407.1 | - | 2 | 457 | 6180242 | ||
BI257596.1 | - | 2 | 281 | |||
BM762761.1 | - | 2 | 488 | |||
BQ932163.1 | - | 3 | 318 | |||
BT019345.1 | - | 1 | 99 | |||
DA568152.1 | - | 2 | 412 |
Gene Symbol: UBE2E4P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110506.1 | 1 | 102 | |||
GenBank mRNA | AF136176.1 | - | 1 | 102 |