Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YWHAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012479.3 | NP_036611.2 | 2 | 407 | ||
GenBank mRNA | AB024334.1 | - | 2 | 359 | ||
AK024230.1 | - | 2 | 371 | |||
AK293505.1 | - | 2 | 371 | |||
AK295029.1 | - | 3 | 286 | |||
BC020963.2 | - | 2 | 381 | 3915246 | ||
CR541904.1 | - | 2 | 190 | |||
CR541925.1 | - | 2 | 190 | |||
DC343104.1 | - | 2 | 435 |