Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281727.1 | NP_001268656.1 | 3, 4 | 323 | ||
NM_001281728.1 | NP_001268657.1 | 4, 5 | 402 | |||
NM_001281729.1 | NP_001268658.1 | 4, 5 | 710 | |||
NM_014183.3 | NP_054902.1 | 3, 4 | 327 | |||
NM_177953.2 | NP_808852.1 | 4, 5 | 406 | |||
NR_104032.1 | 4, 5 | 409 | ||||
GenBank mRNA | AF132750.1 | - | 3, 4 | 307 | ||
AF161511.1 | - | 3, 4 | 298 | |||
AF165516.1 | - | 3, 4 | 280 | |||
AF178431.1 | - | 2, 3 | 252 | |||
AK026864.1 | - | 2, 3 | 263 | |||
AY026513.1 | - | 2, 3 | 252 | |||
BC002481.1 | - | 3, 4 | 305 | 3347555 | ||
BG110161.1 | - | 3, 4 | 312 | |||
BG419592.1 | - | 4, 5 | 378 | |||
BM561321.1 | - | 4, 5 | 680 | |||
BM791453.1 | - | 4, 5 | 388 | |||
BQ639804.1 | - | 4, 5 | 375 | |||
BU163605.1 | - | 346 | ||||
CB149456.1 | - | 3, 4 | 323 |