Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004686.4 | NP_004677.3 | 4 | 693 | ||
XM_017013956.1 | XP_016869445.1 | 4 | 515 | |||
GenBank mRNA | AK294468.1 | - | 4 | 454 | ||
AK316022.1 | - | 4 | 503 | |||
AK316283.1 | - | 4 | 513 | |||
BC130577.1 | - | 4 | 479 | 40146608 | ||
BC130579.1 | - | 4 | 479 | 40146610 | ||
CR749240.1 | - | 4 | 491 |