Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004686.4 | NP_004677.3 | 9, 10 | 1339 | ||
XM_006716414.3 | XP_006716477.2 | 4, 5 | 1705 | |||
XM_011544692.2 | XP_011542994.1 | 6, 7 | 778 | |||
XM_011544693.2 | XP_011542995.1 | 5, 6 | 1240 | |||
XM_017013956.1 | XP_016869445.1 | 8, 9 | 1028 | |||
GenBank mRNA | AF073482.1 | - | 5, 6 | 608 | ||
AK294468.1 | - | 9, 10 | 1100 | |||
AK303762.1 | - | 1, 2 | 162 | |||
AK316022.1 | - | 9, 10 | 1149 | |||
AK316283.1 | - | 9, 10 | 1159 | |||
BC130577.1 | - | 9, 10 | 1125 | 40146608 | ||
BC130579.1 | - | 9, 10 | 1125 | 40146610 | ||
CR749240.1 | - | 9, 10 | 1137 |